Learn More
Cayman Chemical ClozapIn-d4 50ug
SDP
Catalog No. 502967758
Encompass_Preferred
An internal standard for the quantification of clozapine by GC- or LC-MS
Catalog No. 50-296-7758 Supplier Cayman Chemical Supplier No. 1751350 UG
An internal standard for the quantification of clozapine by GC- or LC-MS