Learn More
Cayman Chemical ClozapIn N-oxIde 25mg
SDP
Supplier:  Cayman Chemical 1688225 MG
Encompass_Preferred
A metabolite of clozapine and muscarinic DREADD agonist (EC50s = 16.7, 323, 17.4, 18.3, and 18.7 nM for PASMCs expressing hM1-5D receptors, respectively); prevents action potential firing in cultured hippocampal neurons transiently expressing hM4D receptors; increases glutamate in the nucleus accumbens core and inhibits cue-induced reinstatement of cocaine-seeking behavior in rats transfected with hM3Dq DREADD receptors
Catalog No. 50-298-0011