Learn More
Cayman Chemical Clozapine N-oxide (hydrochloride), Purity: ≥98%, CAS: 2250025-93-3, 10 mg
SDP
Catalog No. 502936094
Encompass_Preferred
A metabolite of clozapine and muscarinic DREADD agonist (EC50s = 16.7, 323, 17.4, 18.3, and 18.7 nM for PASMCs expressing hM1-5D receptors, respectively); prevents action potential firing in cultured hippocampal neurons transiently expressing hM4D receptors; increases glutamate in the nucleus accumbens core and inhibits cue-induced reinstatement of cocaine-seeking behavior in rats transfected with hM3Dq DREADD receptors
Catalog No. 50-293-6094 Supplier Cayman Chemical Supplier No. 2578010 MG
A metabolite of clozapine and muscarinic DREADD agonist (EC50s = 16.7, 323, 17.4, 18.3, and 18.7 nM for PASMCs expressing hM1-5D receptors, respectively); prevents action potential firing in cultured hippocampal neurons transiently expressing hM4D receptors; increases glutamate in the nucleus accumbens core and inhibits cue-induced reinstatement of cocaine-seeking behavior in rats transfected with hM3Dq DREADD receptors