Learn More
Cayman Chemical ClozapIn NoxIdehydrochLR 50mg
SDP
Catalog No. 502936523
Encompass_Preferred
An MCT1 and MPC inhibitor inhibits lactate update in SiHa human cervix carcinoma cells (IC50 50 nM) inhibits proliferation of SiHa cells in the presence of lactate but not glucose inhibits MPC in isolated tumor cell mitochondria increases intratumor lactate accumulation and decreases tumor size in an SiHa mouse xenograft model at 3 mg/kg
Catalog No. 50-293-6523 Supplier Cayman Chemical Supplier No. 2578050 MG
An MCT1 and MPC inhibitor inhibits lactate update in SiHa human cervix carcinoma cells (IC50 50 nM) inhibits proliferation of SiHa cells in the presence of lactate but not glucose inhibits MPC in isolated tumor cell mitochondria increases intratumor lactate accumulation and decreases tumor size in an SiHa mouse xenograft model at 3 mg/kg