Learn More
Medchemexpress LLC Clozapine | 5786-21-0 | 99.8% | C18H19ClN4 | 1 ML
SDP
Catalog No. 5000355237
Encompass_Preferred
Clozapine is an antipsychotic agent used for schizophrenia research. It exhibits high affinity for various neuroreceptors, including potent antagonism of dopamine D2, and inhibition of muscarinic M1 and serotonin 5HT2A receptors. Clozapine is also a potent and selective agonist at the muscarinic M4 receptor.
  • Used for schizophrenia research
  • Acts as an antipsychotic agent
  • Potent antagonist of dopamine D2 receptor
  • Inhibitor of muscarinic M1 and serotonin 5HT2A receptors
  • Selective agonist at muscarinic M4 receptor
Catalog No. 50-003-55237 Supplier Medchemexpress LLC Supplier No. HY1453910MM/1ML
Clozapine is an antipsychotic agent used for schizophrenia research. It exhibits high affinity for various neuroreceptors, including potent antagonism of dopamine D2, and inhibition of muscarinic M1 and serotonin 5HT2A receptors. Clozapine is also a potent and selective agonist at the muscarinic M4 receptor.
  • Used for schizophrenia research
  • Acts as an antipsychotic agent
  • Potent antagonist of dopamine D2 receptor
  • Inhibitor of muscarinic M1 and serotonin 5HT2A receptors
  • Selective agonist at muscarinic M4 receptor