Learn More
Selleck Chemical LLC Clozapine S2459-1g
SDP
Catalog No. 503112621
Encompass_Preferred
Clozapine (HF 1854 LX 100-129) is an atypical antipsychotic drug by acting as a 5-HT antagonist used in the treatment of schizophrenia
Catalog No. 50-311-2621 Supplier Selleck Chemical LLC Supplier No. S24591G
Clozapine (HF 1854 LX 100-129) is an atypical antipsychotic drug by acting as a 5-HT antagonist used in the treatment of schizophrenia