Learn More
Medchemexpress LLC Clozapine-d8 | 1185053-50-2 | MFCD09752113 | 99.7% | 334.87 | C18H11D8ClN4 | 5 MG
SDP
Catalog No. 5000221657
Encompass_Preferred
Clozapine-d8 is the deuterium-labeled isotopologue of the antipsychotic clozapine, provided as a high-purity analytical standard for research use. It is intended as an internal standard for quantitative bioanalytical methods and preclinical studies where accurate measurement of clozapine is required.
  • Deuterium-labeled internal standard for accurate quantitation.
  • Suitable for LC-MS/MS and GC-MS analytical methods.
  • High purity (≈99.7%).
  • Provides improved assay precision and recovery.
  • Supplied in small quantities appropriate for analytical standards.
Catalog No. 50-002-21657 Supplier Medchemexpress LLC Supplier No. HY14539S5MG
Clozapine-d8 is the deuterium-labeled isotopologue of the antipsychotic clozapine, provided as a high-purity analytical standard for research use. It is intended as an internal standard for quantitative bioanalytical methods and preclinical studies where accurate measurement of clozapine is required.
  • Deuterium-labeled internal standard for accurate quantitation.
  • Suitable for LC-MS/MS and GC-MS analytical methods.
  • High purity (≈99.7%).
  • Provides improved assay precision and recovery.
  • Supplied in small quantities appropriate for analytical standards.