Learn More
TARGETMOL CHEMICALS INC CLOZAPINE N-OXIDE 25MG
SDP
Catalog No. 502576759
Encompass_Preferred
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Clozapine N-oxide is a major metabolite of clozapine that can be detected in the earliest excretion time interval after clozapine administration [1]. It is produced by the action of cytochrome P450 (CYP) isoforms CYP1A2 and CYP3A4 whose activities in turn can be affected by drugs and disease [2]. Clozapine N-oxide is normally biologically inactive. However it can be used in designer receptor exclusively activated by designer drugs also known as DREADDs technology to modulate synthetic receptors in vivo [3]. purity: 99%
Catalog No. 50-257-6759 Supplier TARGETMOL CHEMICALS INC Supplier No. T449425MG
Also available in 2 mg 5 mg 10 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Clozapine N-oxide is a major metabolite of clozapine that can be detected in the earliest excretion time interval after clozapine administration [1]. It is produced by the action of cytochrome P450 (CYP) isoforms CYP1A2 and CYP3A4 whose activities in turn can be affected by drugs and disease [2]. Clozapine N-oxide is normally biologically inactive. However it can be used in designer receptor exclusively activated by designer drugs also known as DREADDs technology to modulate synthetic receptors in vivo [3]. purity: 99%