Learn More
Medchemexpress LLC Clozapine N-oxide | 34233-69-7 | 99.9% | 342.82 | 100 MG
SDP
Catalog No. 5000384757
Encompass_Preferred
Clozapine N-oxide is a major metabolite of clozapine and a human muscarinic designer receptors (DREADDs) agonist. It activates the DREADD receptor hM3Dq and hM4Di but cannot cross the blood-brain barrier. Its parent compound, clozapine, is a potent dopamine antagonist and a potent and selective muscarinic M4 receptor agonist. For research use only.
  • DREADD activator
  • Metabolite of clozapine
  • Activates hM3Dq and hM4Di DREADD receptors
  • Cannot cross the blood-brain barrier
  • Solid appearance, light yellow to yellow color
Catalog No. 50-003-84757 Supplier Medchemexpress LLC Supplier No. HY17366100MG
Clozapine N-oxide is a major metabolite of clozapine and a human muscarinic designer receptors (DREADDs) agonist. It activates the DREADD receptor hM3Dq and hM4Di but cannot cross the blood-brain barrier. Its parent compound, clozapine, is a potent dopamine antagonist and a potent and selective muscarinic M4 receptor agonist. For research use only.
  • DREADD activator
  • Metabolite of clozapine
  • Activates hM3Dq and hM4Di DREADD receptors
  • Cannot cross the blood-brain barrier
  • Solid appearance, light yellow to yellow color