Learn More
Medchemexpress LLC Clozapine N-oxide | 34233-69-7 | 99.9% | 342.82 | 25 MG
SDP
Catalog No. 5000384124
Encompass_Preferred
Clozapine N-oxide is a major metabolite of Clozapine, acting as an agonist for human muscarinic designer receptors (DREADDs). It activates the hM3Dq and hM4Di DREADD receptors, but it cannot cross the blood-brain barrier. Its parent compound, Clozapine, is a potent dopamine antagonist and a selective muscarinic M4 receptor agonist.
  • Activates DREADD receptors hM3Dq and hM4Di
  • Cannot cross the blood-brain barrier
  • Metabolite of Clozapine
  • Solid appearance
  • Light yellow to yellow color
Catalog No. 50-003-84124 Supplier Medchemexpress LLC Supplier No. 25MGHY17366
Clozapine N-oxide is a major metabolite of Clozapine, acting as an agonist for human muscarinic designer receptors (DREADDs). It activates the hM3Dq and hM4Di DREADD receptors, but it cannot cross the blood-brain barrier. Its parent compound, Clozapine, is a potent dopamine antagonist and a selective muscarinic M4 receptor agonist.
  • Activates DREADD receptors hM3Dq and hM4Di
  • Cannot cross the blood-brain barrier
  • Metabolite of Clozapine
  • Solid appearance
  • Light yellow to yellow color