Learn More
Medchemexpress LLC Clozapine N-oxide | 34233-69-7 | 99.9% | 342.82 | 50 MG
SDP
Catalog No. 5000385198
Encompass_Preferred
Clozapine N-oxide is a major metabolite of Clozapine and acts as an agonist for human muscarinic designer receptors (DREADDs). It activates the DREADD receptors hM3Dq and hM4Di. Clozapine N-oxide cannot cross the blood-brain barrier. It undergoes reverse-metabolism to its parent compound clozapine, an atypical antipsychotic.
  • Activates DREADD receptors hM3Dq and hM4Di
  • Cannot cross the blood-brain barrier
  • Undergoes reverse-metabolism to clozapine
  • Suitable for laboratory centrifugation and storage
Catalog No. 50-003-85198 Supplier Medchemexpress LLC Supplier No. HY1736650MG
Clozapine N-oxide is a major metabolite of Clozapine and acts as an agonist for human muscarinic designer receptors (DREADDs). It activates the DREADD receptors hM3Dq and hM4Di. Clozapine N-oxide cannot cross the blood-brain barrier. It undergoes reverse-metabolism to its parent compound clozapine, an atypical antipsychotic.
  • Activates DREADD receptors hM3Dq and hM4Di
  • Cannot cross the blood-brain barrier
  • Undergoes reverse-metabolism to clozapine
  • Suitable for laboratory centrifugation and storage