Learn More
Medchemexpress LLC Clozapine N-oxide | 34233-69-7 | 99.98% | 342.82 | 50 MG
SDP
Catalog No. 5000384224
Encompass_Preferred
Clozapine N-oxide is a major metabolite of Clozapine and acts as a human muscarinic designer receptors (DREADDs) agonist. It activates the DREADD receptor hM3Dq and hM4Di but cannot cross the blood-brain barrier. Clozapine, its parent compound, is a potent dopamine antagonist and a selective muscarinic M4 receptor agonist.
  • Activates DREADD receptor hM3Dq and hM4Di.
  • For research use only.
  • Provides details on solubility and stability in various solvents for both in vitro and in vivo applications.
  • Cited in numerous top publications from various scientific journals.
Catalog No. 50-003-84224 Supplier Medchemexpress LLC Supplier No. HY17366A50MG
Clozapine N-oxide is a major metabolite of Clozapine and acts as a human muscarinic designer receptors (DREADDs) agonist. It activates the DREADD receptor hM3Dq and hM4Di but cannot cross the blood-brain barrier. Clozapine, its parent compound, is a potent dopamine antagonist and a selective muscarinic M4 receptor agonist.
  • Activates DREADD receptor hM3Dq and hM4Di.
  • For research use only.
  • Provides details on solubility and stability in various solvents for both in vitro and in vivo applications.
  • Cited in numerous top publications from various scientific journals.