Learn More
Apexbio Technology LLC Clozapine N-oxide (CNO) 34233-69-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062817
Encompass_Preferred
Clozapine N-oxide (CNO CAS 34233-69-7) is a metabolic derivative of the atypical antipsychotic drug clozapine primarily known for its applications as a chemogenetic actuator Biologically inert in typical mammalian systems CNO selectively activates engineered muscarinic receptors such as the M3-designer receptors exclusively activated by designer drugs (DREADDs) Additionally CNO influences receptor expression levels exposure in cortical neuron cultures reduces 5-HT2 receptor density significantly Due to its specificity CNO is extensively utilized in neuroscience research to modulate neuronal activity non-invasively through chemogenetic approaches
Catalog No. 50-000-62817 Supplier Apexbio Technology LLC Supplier No. A33175.1
Clozapine N-oxide (CNO CAS 34233-69-7) is a metabolic derivative of the atypical antipsychotic drug clozapine primarily known for its applications as a chemogenetic actuator Biologically inert in typical mammalian systems CNO selectively activates engineered muscarinic receptors such as the M3-designer receptors exclusively activated by designer drugs (DREADDs) Additionally CNO influences receptor expression levels exposure in cortical neuron cultures reduces 5-HT2 receptor density significantly Due to its specificity CNO is extensively utilized in neuroscience research to modulate neuronal activity non-invasively through chemogenetic approaches