Learn More
TARGETMOL CHEMICALS INC Clozapine N-oxide dihydrochlor
SDP
Catalog No. 502878209
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Clozapine N-oxide dihydrochloride, a derivative of Clozapine and an agonist of human muscarinic design receptors (DREADDs), crosses the blood-brain barrier and activates DREADD receptors hM3Dq and hM4Di. Purity 98.11%
Catalog No. 50-287-8209 Supplier TARGETMOL CHEMICALS INC Supplier No. T7211225MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Clozapine N-oxide dihydrochloride, a derivative of Clozapine and an agonist of human muscarinic design receptors (DREADDs), crosses the blood-brain barrier and activates DREADD receptors hM3Dq and hM4Di. Purity 98.11%