Learn More
Medchemexpress LLC Clozapine N-oxide (dihydrochloride) | 2250025-93-3 | 99.7% | 415.74 | 100 MG
SDP
Catalog No. 5000384623
Encompass_Preferred
Clozapine N-oxide dihydrochloride is a major metabolite of Clozapine and acts as an agonist for human muscarinic designer receptors (DREADDs). It activates the DREADD receptors hM3Dq and hM4Di. This compound cannot cross the blood-brain barrier. Clozapine, its parent compound, is a potent dopamine antagonist and a selective muscarinic M4 receptor agonist with an EC50 of 11 nM.
  • Major metabolite of Clozapine
  • Activates human muscarinic designer receptors (DREADDs) hM3Dq and hM4Di
  • Cannot cross the blood-brain barrier
  • Clozapine is a potent dopamine antagonist and selective muscarinic M4 receptor agonist
  • Appearance: Solid
  • Color: Light yellow to yellow
Catalog No. 50-003-84623 Supplier Medchemexpress LLC Supplier No. HY17366A100MG
Clozapine N-oxide dihydrochloride is a major metabolite of Clozapine and acts as an agonist for human muscarinic designer receptors (DREADDs). It activates the DREADD receptors hM3Dq and hM4Di. This compound cannot cross the blood-brain barrier. Clozapine, its parent compound, is a potent dopamine antagonist and a selective muscarinic M4 receptor agonist with an EC50 of 11 nM.
  • Major metabolite of Clozapine
  • Activates human muscarinic designer receptors (DREADDs) hM3Dq and hM4Di
  • Cannot cross the blood-brain barrier
  • Clozapine is a potent dopamine antagonist and selective muscarinic M4 receptor agonist
  • Appearance: Solid
  • Color: Light yellow to yellow