Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Clozapine N-oxide dihydrochloride | 2250025-93-3 | MFCD00210190 | 10 MG
SDP

Catalog No. 5000298718
Encompass_Preferred

Clozapine N-oxide dihydrochloride is the dihydrochloride salt of clozapine N-oxide, a metabolite of clozapine commonly used as a selective agonist for designer receptors exclusively activated by designer drugs (DREADDs). The compound is provided as a research reagent for neuroscience and pharmacology studies; the salt form improves aqueous solubility compared with the free base and is intended for experimental use in vitro and in vivo.

  • Activates DREADD receptors hM3Dq and hM4Di.
  • Dihydrochloride salt with improved aqueous solubility.
  • Molecular formula: C18H21Cl3N4O.
  • Molecular weight: 415.74.
  • CAS number: 2250025-93-3.
  • Intended for research applications in neuroscience and pharmacology.
  • Provided in small research quantities for experimental use.

Catalog No. 50-002-98718 Supplier Medchemexpress LLC Supplier No. HY17366A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Clozapine N-oxide dihydrochloride is the dihydrochloride salt of clozapine N-oxide, a metabolite of clozapine commonly used as a selective agonist for designer receptors exclusively activated by designer drugs (DREADDs). The compound is provided as a research reagent for neuroscience and pharmacology studies; the salt form improves aqueous solubility compared with the free base and is intended for experimental use in vitro and in vivo.

  • Activates DREADD receptors hM3Dq and hM4Di.
  • Dihydrochloride salt with improved aqueous solubility.
  • Molecular formula: C18H21Cl3N4O.
  • Molecular weight: 415.74.
  • CAS number: 2250025-93-3.
  • Intended for research applications in neuroscience and pharmacology.
  • Provided in small research quantities for experimental use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.