Learn More
Medchemexpress LLC Clozapine (Standard) | 5786-21-0 | 99.98% | C₁₈H₁₉ClN₄ | 100 MG
SDP
Catalog No. 5000354192
Encompass_Preferred
Clozapine (Standard) is the analytical standard of Clozapine, intended for research and analytical applications. It is an antipsychotic used for the research of schizophrenia. Clozapine exhibits high affinity for various neuroreceptors, acting as a potent antagonist of dopamine D2 and inhibiting muscarinic M1 and serotonin 5HT2A receptors. It is also a potent and selective agonist at the muscarinic M4 receptor.
  • Intended for research and analytical applications
  • Analytical standard of Clozapine
  • Used for schizophrenia research
  • Potent antagonist of dopamine D2
  • Inhibits muscarinic M1 and serotonin 5HT2A receptors
  • Potent and selective agonist at the muscarinic M4 receptor
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS
Catalog No. 50-003-54192 Supplier Medchemexpress LLC Supplier No. HY14539R100MG
Clozapine (Standard) is the analytical standard of Clozapine, intended for research and analytical applications. It is an antipsychotic used for the research of schizophrenia. Clozapine exhibits high affinity for various neuroreceptors, acting as a potent antagonist of dopamine D2 and inhibiting muscarinic M1 and serotonin 5HT2A receptors. It is also a potent and selective agonist at the muscarinic M4 receptor.
  • Intended for research and analytical applications
  • Analytical standard of Clozapine
  • Used for schizophrenia research
  • Potent antagonist of dopamine D2
  • Inhibits muscarinic M1 and serotonin 5HT2A receptors
  • Potent and selective agonist at the muscarinic M4 receptor
  • Used in qualitative, quantitative and methodological research experiments in HPLC, GC and MS