Learn More
Medchemexpress LLC Clozapine (Standard) | 5786-21-0 | 99.98% | C₁₈H₁₉ClN₄ | 50 MG
SDP
Catalog No. 5000354193
Encompass_Preferred
Clozapine (Standard) is the analytical standard for Clozapine, intended for research and analytical applications. It is an antipsychotic used in schizophrenia research, exhibiting high affinity for various neuroreceptors. It acts as a potent antagonist of dopamine D2 and inhibits muscarinic M1 and serotonin 5HT2A receptors, also functioning as a potent and selective agonist at the muscarinic M4 receptor.
  • Analytical standard for research and analytical applications
  • Used in schizophrenia research
  • High affinity for various neuroreceptors
  • Potent antagonist of dopamine D2 with a Ki of 75 nM
  • Inhibits muscarinic M1 receptor and serotonin 5HT2A receptor with Kis of 9.5 nM and 4 nM
  • Potent and selective agonist at the muscarinic M4 receptor (EC50=11 nM)
  • Commonly used in qualitative, quantitative, and methodological research experiments
Catalog No. 50-003-54193 Supplier Medchemexpress LLC Supplier No. HY14539R50MG
Clozapine (Standard) is the analytical standard for Clozapine, intended for research and analytical applications. It is an antipsychotic used in schizophrenia research, exhibiting high affinity for various neuroreceptors. It acts as a potent antagonist of dopamine D2 and inhibits muscarinic M1 and serotonin 5HT2A receptors, also functioning as a potent and selective agonist at the muscarinic M4 receptor.
  • Analytical standard for research and analytical applications
  • Used in schizophrenia research
  • High affinity for various neuroreceptors
  • Potent antagonist of dopamine D2 with a Ki of 75 nM
  • Inhibits muscarinic M1 receptor and serotonin 5HT2A receptor with Kis of 9.5 nM and 4 nM
  • Potent and selective agonist at the muscarinic M4 receptor (EC50=11 nM)
  • Commonly used in qualitative, quantitative, and methodological research experiments