Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLP ICP Interference Check Standard for ILM 4.0, 16 Analytes, Ultra Scientific

Catalog No. USICM443
Encompass
Change view
Click to view available options
Quantity:
125 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
USICM443 125 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. USICM443 Supplier Agilent Technologies Supplier No. ICM443
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Specifications

Chemical Name or Material CLP ICP Interference Check Standard
CAS Max % 97.92, 2.0, 0.01, 0.0, 0.0
CAS 7440-36-0, 7440-38-2, 7440-39-3, 7697-37-2, 7732-18-5
Physical Form Liquid
Quantity 125 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.