Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLP Volatiles Mixture 4 (For EPA CLP Methods)

Catalog No. USCLP154
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-CLP-154 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-CLP-154 Supplier Agilent Technologies Supplier No. CLP1541
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Specifications

Chemical Name or Material CLP Volatiles Mixture 4
Packaging Ampule
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) Each component at 2000μg/mL in Methanol:
Acetone
2-Butanone
Carbon Disulfide
2-Hexanone
4-Methyl-2-pentanone
Styrene
o-Xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.