Learn More
Aviva Systems Biology Corp CLP1 Antibody - C-terminal region (ARP64201_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP64201_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLP1. It was validated on Western Blot. . Alternate names: HEAB, hClp1ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-288-201