Learn More
Medchemexpress LLC HY-103023 10mg Medchemexpress, CLP290 CAS:1181083-81-7 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY10302310MG
Encompass_Preferred
Medchemexpress, HY-103023 10mg CLP290 CAS:1181083-81-7 CLP290 is an orally available activator of the neuron-specific K+-Cl− cotransporter KCC2, displays potential for treatment of a wide range of neurological and psychiatric indications. CLP290 can significantly lower blood arginine-vasopressin (AVP) and glucose levels in STZ rats. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8017