Learn More
Grainger Clps Bulk Ctr Black Solid 32 in
Catalog No. 502752770
Encompass_Preferred
The Grainger 14L050 is a Westward brand 1/4-inch drive click-type torque wrench with a range of 10 to 50 foot pounds. It has a reversible ratchet head and comes with a molded storage case. The torque wrench has an accuracy of ±4% clockwise and has a chrome vanadium steel construction with a satin chrome finish. It measures 15-1/2 inches long and weighs 1.8 pounds. This torque wrench is designed for accurate tightening and loosening of threaded fasteners.
Catalog No. 50-275-2770 Supplier Grainger Supplier No. 14L050
The Grainger 14L050 is a Westward brand 1/4-inch drive click-type torque wrench with a range of 10 to 50 foot pounds. It has a reversible ratchet head and comes with a molded storage case. The torque wrench has an accuracy of ±4% clockwise and has a chrome vanadium steel construction with a satin chrome finish. It measures 15-1/2 inches long and weighs 1.8 pounds. This torque wrench is designed for accurate tightening and loosening of threaded fasteners.