Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spex CertiPrep™ CLPV Series Volatile Analyte Mix A
SDP

Catalog No. CLPVAPK Shop All Spex CertiPrep Products
Change view
Click to view available options
Quantity:
4/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CLPVAPK 4/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CLPVAPK Supplier Spex CertiPrep™ Supplier No. CLPVAPK

missing translation for 'validMainframeMsg'

Analytical standards for drinking water and raw source water

  • Accredited by A2LA to ISO/IEC 17025:2017 and ISO 17034:2016
  • Manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
  • For US EPA 502.1, 502.2, 524, 624, 8260, and CLP Series Methods

Specifications

Chemical Name or Material 1,1,2,2-Tetrachloroethane
Packaging Bottle
CAS 79-34-5
Concentration 200 μg/mL
Color Clear
Melting Point Undetermined
Boiling Point 64°C (147°F)
Solution Type Standard Solution
Type Organic Pollutant Standard
Industry Type Environmental Testing
Quantity 4/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.