Learn More
GA International Cryogenic Lamination Tape 0.75" x 200' / 19.1mm x 60m Transparent From -196°C to +93°C (-321°F to +199°F)
Catalog No. NC0750724
Encompass_Preferred
This ultra-thin cryo laminating tape provides excellent protection against most chemicals and reagents. It has also been designed to withstand long-term storage in liquid phase and vapor phase liquid nitrogen and ultra-low temperature freezers (as low as -196°C) when applied on cryo vials and tubes. Resistant to dry ice, multiple freeze-thaw cycles, and other harsh laboratory environments.
Catalog No. NC0750724 Supplier GA International Supplier No. CLAM19C3200
This ultra-thin cryo laminating tape provides excellent protection against most chemicals and reagents. It has also been designed to withstand long-term storage in liquid phase and vapor phase liquid nitrogen and ultra-low temperature freezers (as low as -196°C) when applied on cryo vials and tubes. Resistant to dry ice, multiple freeze-thaw cycles, and other harsh laboratory environments.