Learn More
Waters Corp Waters Vial, LCGC Certified, Clear Glass, 1 mL, Snap Neck
Catalog No. 50818721 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Certified vials are rigorously tested for cleanliness by HPLC and adhere to the industry’s most stringent dimensional tolerances. These LCGC certified vials are designed with a snap neck for tool-free assembly, featuring a plastic cap that creates a secure seal. Constructed from Type 1, 33-Expansion Borosilicate Glass, they offer superior chemical inertness for reliable analytical results.
  • Tested for cleanliness by HPLC.
  • Held to tightest dimensional tolerances in the industry.
  • Snap neck design for easy assembly without tools.
  • Made from Type 1, 33-Expansion Borosilicate Glass for chemical inertness.
  • Each package contains 250 vials.
  • Includes vial, cap, and septum.
  • Suitable for sample preparation.
  • LCGC certified.
Catalog No. 50-818-721 Supplier Waters Corp Supplier No. WAT025054C
Waters Certified vials are rigorously tested for cleanliness by HPLC and adhere to the industry’s most stringent dimensional tolerances. These LCGC certified vials are designed with a snap neck for tool-free assembly, featuring a plastic cap that creates a secure seal. Constructed from Type 1, 33-Expansion Borosilicate Glass, they offer superior chemical inertness for reliable analytical results.
  • Tested for cleanliness by HPLC.
  • Held to tightest dimensional tolerances in the industry.
  • Snap neck design for easy assembly without tools.
  • Made from Type 1, 33-Expansion Borosilicate Glass for chemical inertness.
  • Each package contains 250 vials.
  • Includes vial, cap, and septum.
  • Suitable for sample preparation.
  • LCGC certified.