Learn More
Electron Microscopy Sciences StainTray™ Clear Lid Only for 20 Slide Stain Tray
SDP
Catalog No. 501899567
Encompass_Preferred
A clear cover, allowing for visual examination, made from PETG with temperature a range of -20ºC to 60ºC.
Capacity: 20 slides
Catalog No. 50-189-9567 Supplier Electron Microscopy Sciences Supplier No. 71399C
A clear cover, allowing for visual examination, made from PETG with temperature a range of -20ºC to 60ºC.
Capacity: 20 slides