Learn More
Labsource Inc Clear Weld Heat Seal Mark 2, Permanent Heat Seal, Optically Clear Polymer Film, 500 m x 115 mm
SDP
Catalog No. NC3618483
Encompass
The Clear Weld Heat Seal Mark 2 is an optically clear, permanent sealing film specifically designed for scientific applications. It offers robust protection for polypropylene plates and ensures maximum seal integrity, making it ideal for critical assays such as PCR and qPCR. With dimensions of 500 m x 115 mm, it is autoclavable and resistant to solvents, ensuring reliability for both low-temperature storage and high-temperature incubations.
Catalog No. NC3618483 Supplier Labsource Inc Supplier No. P98646A
The Clear Weld Heat Seal Mark 2 is an optically clear, permanent sealing film specifically designed for scientific applications. It offers robust protection for polypropylene plates and ensures maximum seal integrity, making it ideal for critical assays such as PCR and qPCR. With dimensions of 500 m x 115 mm, it is autoclavable and resistant to solvents, ensuring reliability for both low-temperature storage and high-temperature incubations.