Learn More
Bellco Glass, Inc. Clr PolyCrimp Seal, 11mm 10mil PTFE
SDP
Catalog No. 501945415
Encompass_Preferred
Larger opening provides increased target area for needle penetration. MADE IN THE USA. Size: 11mm Septa material: ptfe silicone Seal color: silver and clear
Catalog No. 50-194-5415 Supplier Bellco Glass, Inc. Supplier No. 207005400
Larger opening provides increased target area for needle penetration. MADE IN THE USA. Size: 11mm Septa material: ptfe silicone Seal color: silver and clear