Learn More
New England Biolabs, Inc. Color Prestained Protein Standard, Broad Range (10-250 kDa) – 150 gel lanes
SDP
Catalog No. NC1663304
Encompass
The Color Prestained Protein Standard, Broad Range is a mixture of highly pure, recombinant, prestained proteins, covalently coupled with a blue chromophore, and two reference bands (one orange and one green at 72 kDa and 26 kDa, respectively), that resolves into 11 sharp bands when electrophoresed.
Catalog No. NC1663304 Supplier New England Biolabs, Inc. Supplier No. P7719S
The Color Prestained Protein Standard, Broad Range is a mixture of highly pure, recombinant, prestained proteins, covalently coupled with a blue chromophore, and two reference bands (one orange and one green at 72 kDa and 26 kDa, respectively), that resolves into 11 sharp bands when electrophoresed.