Learn More
MSC CLR Pro Bio-Enzymatic Drain Maintainer, Live Microbial, 5 gal
Catalog No. 502820009
Encompass
CLR PRO® Bio-Enzymatic Drain Maintainer is formulated as a live microbial solution to prevent the buildup of fats, oils, and grease (FOG). It is suitable for regular maintenance use in drains, grease traps, and septic systems. The product is also certified by NSF, EPA Safer Choice, and is biodegradable and LEED Certified.
  • Live microbial formula
  • Prevents FOG buildup
  • Treats drains, grease traps, and septic systems
  • Designed for regular maintenance use
  • All-purpose cleaner and degreaser
  • Liquid form
  • Odor-free
  • Container size: 5 gal pail
  • Disinfectant: No
  • Standards: NSF; EPA Safer Choice; Biodegradable; LEED Certified
  • Formula type: Probiotic
  • Solution type: Ready to use; water-based
  • Non-acidic
Catalog No. 50-282-0009 Supplier MSC Supplier No. 28514016
CLR PRO® Bio-Enzymatic Drain Maintainer is formulated as a live microbial solution to prevent the buildup of fats, oils, and grease (FOG). It is suitable for regular maintenance use in drains, grease traps, and septic systems. The product is also certified by NSF, EPA Safer Choice, and is biodegradable and LEED Certified.
  • Live microbial formula
  • Prevents FOG buildup
  • Treats drains, grease traps, and septic systems
  • Designed for regular maintenance use
  • All-purpose cleaner and degreaser
  • Liquid form
  • Odor-free
  • Container size: 5 gal pail
  • Disinfectant: No
  • Standards: NSF; EPA Safer Choice; Biodegradable; LEED Certified
  • Formula type: Probiotic
  • Solution type: Ready to use; water-based
  • Non-acidic