Learn More
MSC CLR Pro Heavy Duty Cleaner Degreaser, Biodegradable, 1 gal
Catalog No. 502867532
Encompass
CLR Pro Heavy Duty Cleaner & Degreaser is a liquid, water-based solution designed to quickly lift out oil, grease, and tar from a variety of surfaces. This ready-to-use, all-purpose formula comes in a 1-gallon jug with a handle.
  • Cuts through oil, grease, cutting fluids, and residue on parts, machinery, and heavy equipment.
  • Used for surface preparation prior to welding, plating, and painting.
  • Safe to use in floor scrubbers, parts washers, dip tanks, pressure washers, and carpet extractors.
  • Water-based and biodegradable; will not harm surrounding vegetation.
  • Contains no acids, abrasive, or petroleum solvents.
  • NSF certified, approved for use in food processing, food storage, and food service facilities.
  • Removes grease, oil, and food residue from appliances, vents, traps, grill hoods, and trash receptacles.
Catalog No. 50-286-7532 Supplier MSC Supplier No. 28514057
CLR Pro Heavy Duty Cleaner & Degreaser is a liquid, water-based solution designed to quickly lift out oil, grease, and tar from a variety of surfaces. This ready-to-use, all-purpose formula comes in a 1-gallon jug with a handle.
  • Cuts through oil, grease, cutting fluids, and residue on parts, machinery, and heavy equipment.
  • Used for surface preparation prior to welding, plating, and painting.
  • Safe to use in floor scrubbers, parts washers, dip tanks, pressure washers, and carpet extractors.
  • Water-based and biodegradable; will not harm surrounding vegetation.
  • Contains no acids, abrasive, or petroleum solvents.
  • NSF certified, approved for use in food processing, food storage, and food service facilities.
  • Removes grease, oil, and food residue from appliances, vents, traps, grill hoods, and trash receptacles.