Learn More
Tricor Braun 32 oz Clear Glass Boston Round Bottle
Catalog No. NC1673365
Encompass_Preferred
This Boston Round style of bottle was traditionally used in the pharmaceutical and chemical industries. But the graceful shape will work well for a wide variety of products. The bottle is flint (clear) in color and has a 33-400 continuous thread neck finish. Your selection of closure - standard continuous thread, flip top, child resistant, tamper evident, sprayer, etc - will provide any easy way to expand your product line.
Catalog No. NC1673365 Supplier Tricor Braun Supplier No. 097704
This Boston Round style of bottle was traditionally used in the pharmaceutical and chemical industries. But the graceful shape will work well for a wide variety of products. The bottle is flint (clear) in color and has a 33-400 continuous thread neck finish. Your selection of closure - standard continuous thread, flip top, child resistant, tamper evident, sprayer, etc - will provide any easy way to expand your product line.