Learn More
Electron Microscopy Sciences Press-To-Seal Clear Silicone Sheet, 1.6 mm thick
SDP
Catalog No. 5034694
Encompass_Preferred
Isolate specimens using press-to-seal silicone isolators with removable hydrophobic barriers. Silicone isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides. Without Adhesive.
Thickness: 1.6 mm
Catalog No. 50-346-94 Supplier Electron Microscopy Sciences Supplier No. 7033832
Isolate specimens using press-to-seal silicone isolators with removable hydrophobic barriers. Silicone isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides. Without Adhesive.
Thickness: 1.6 mm