Learn More
Electron Microscopy Sciences Press-To-Seal Clear Silicone Sheet, 0.25 mm thick
SDP
Supplier:  Electron Microscopy Sciences 7033879
Encompass_Preferred
Isolate specimens using press-to-seal silicone isolators with removable hydrophobic barriers. Silicone isolators allow researchers to isolate specimens using removable hydrophobic barriers. They may be used to isolate cells grown in culture dishes or separate multiple specimens affixed to microscope slides. Two Sides Adhesive.
Thickness: 0.25 mm
Catalog No. 50-346-80