Learn More
Bellco Glass, Inc. Clr Snap Cap W/PTFE/Sil/PTFE 11mm
SDP
Catalog No. 501945402
Encompass_Preferred
Used for snap thread vials. Caps may be snapped on and off by hand which eliminated the need for a metal crimping and decapping tool. MADE IN THE USA. Size: 11mm Septa material: ptfe
Catalog No. 50-194-5402 Supplier Bellco Glass, Inc. Supplier No. 207003500
Used for snap thread vials. Caps may be snapped on and off by hand which eliminated the need for a metal crimping and decapping tool. MADE IN THE USA. Size: 11mm Septa material: ptfe