Learn More
Aqua Solutions CLR STND 50 500ML
SDP
Catalog No. NC9761148
Encompass_Preferred
Color Standard 50 Units (apha) Prepared To Astm Method D 1209
Catalog No. NC9761148 Supplier Aqua Solutions Supplier No. 2188500ML
Color Standard 50 Units (apha) Prepared To Astm Method D 1209