Learn More
Grainger Cls 2 Trnsfrmr 10VA Enclosed Foot
Catalog No. 502749871
Encompass_Preferred
The Edwards Signaling Class 2 Transformer is a 20 VA, 14 volt AC transformer that steps down 120 volt AC to a lower 14 volt AC output. It has a maximum load rating of 1.4 amps and is intended for use with fire alarm, nurse call, and other low voltage signaling systems. This transformer features a resettable thermal protector, a steel enclosure, and mounting feet for secure installation. With CSA and UL listings, it meets National Electrical Code requirements for use in commercial signaling applications.
Catalog No. 50-274-9871 Supplier Grainger Supplier No. 14W013
The Edwards Signaling Class 2 Transformer is a 20 VA, 14 volt AC transformer that steps down 120 volt AC to a lower 14 volt AC output. It has a maximum load rating of 1.4 amps and is intended for use with fire alarm, nurse call, and other low voltage signaling systems. This transformer features a resettable thermal protector, a steel enclosure, and mounting feet for secure installation. With CSA and UL listings, it meets National Electrical Code requirements for use in commercial signaling applications.