Learn More
Grainger Cls 2 Trnsfrmr 15VA Enclosed Foot
Catalog No. 502750327
Encompass_Preferred
The Edwards Signaling 10VA Class 2 Transformer is a UL Listed Class 2 transformer that steps down 120VAC to 24VAC. It has a 10VA power rating and can support up to 14VA of load. The transformer has an input voltage of 120VAC, 60Hz and an output voltage of 24VAC at 417mA. It features an internal thermal fuse and overcurrent protection. The dimensions are 2-3/4" x 2-1/4" x 2-3/8" and it weighs 1 lb. The 10VA transformer can be used for doorbells, nurse call systems, security systems, fire alarm systems and more. It is ETL Listed to UL Standard 5085-1 and CSA Standard C22.2.
Catalog No. 50-275-0327 Supplier Grainger Supplier No. 14W015
The Edwards Signaling 10VA Class 2 Transformer is a UL Listed Class 2 transformer that steps down 120VAC to 24VAC. It has a 10VA power rating and can support up to 14VA of load. The transformer has an input voltage of 120VAC, 60Hz and an output voltage of 24VAC at 417mA. It features an internal thermal fuse and overcurrent protection. The dimensions are 2-3/4" x 2-1/4" x 2-3/8" and it weighs 1 lb. The 10VA transformer can be used for doorbells, nurse call systems, security systems, fire alarm systems and more. It is ETL Listed to UL Standard 5085-1 and CSA Standard C22.2.