Learn More
Grainger Cls 2 Trnsfrmr 20VA Enclosed Foot
Catalog No. 502749394
Encompass_Preferred
The Dayton Class 2 Transformer is a 20 VA, 120V primary to 4V secondary transformer made by Dayton Electric Manufacturing. It has an aluminum enclosure and features two 4V secondary windings that can be connected in parallel for a 4V output at higher current. The transformer can supply up to 5 amps continuously and has a maximum temperature rise of 115C. It can be mounted on a wall or DIN rail and complies with UL and CSA standards. The transformer measures 3.5" x 3" x 3.25" and weighs 2.5 lbs.
Catalog No. 50-274-9394 Supplier Grainger Supplier No. 14W014
The Dayton Class 2 Transformer is a 20 VA, 120V primary to 4V secondary transformer made by Dayton Electric Manufacturing. It has an aluminum enclosure and features two 4V secondary windings that can be connected in parallel for a 4V output at higher current. The transformer can supply up to 5 amps continuously and has a maximum temperature rise of 115C. It can be mounted on a wall or DIN rail and complies with UL and CSA standards. The transformer measures 3.5" x 3" x 3.25" and weighs 2.5 lbs.