Learn More
Grainger Cls 2 Trnsfrmr 30VA Enclosed Foot
Catalog No. 502750330
Encompass_Preferred
The Edwards Signaling 30VA Class 2 Transformer is a UL Listed Class 2 transformer that steps down 120VAC to 24VAC. It has a maximum load rating of 30VA and provides short circuit protection. This transformer can be used for doorbells, chimes, nurse call systems, security systems, fire alarm systems, and other low voltage control applications. The transformer has an all-steel construction with a scratch resistant powder coated finish, measures 5-1/4" H x 4-3/8" W x 3-3/8" D, and weighs 4 lbs. It comes with a 5 year warranty.
Catalog No. 50-275-0330 Supplier Grainger Supplier No. 14W016
The Edwards Signaling 30VA Class 2 Transformer is a UL Listed Class 2 transformer that steps down 120VAC to 24VAC. It has a maximum load rating of 30VA and provides short circuit protection. This transformer can be used for doorbells, chimes, nurse call systems, security systems, fire alarm systems, and other low voltage control applications. The transformer has an all-steel construction with a scratch resistant powder coated finish, measures 5-1/4" H x 4-3/8" W x 3-3/8" D, and weighs 4 lbs. It comes with a 5 year warranty.