Learn More
Grainger Cls 2 Trnsfrmr 40VA Enclosed Foot
Catalog No. 502750168
Encompass_Preferred
The Edwards Signaling Class 2 Transformer is a 120V AC input, 14V AC output transformer with a maximum load of 40VA. It has an electrostatic shield between primary and secondary windings to minimize line noise interference. This general purpose transformer steps down 120V AC to 14V AC for powering low voltage devices. It has a steel enclosure with knockouts and is UL listed. The transformer measures 5 x 4 x 3 inches and weighs 4 pounds.
Catalog No. 50-275-0168 Supplier Grainger Supplier No. 14W017
The Edwards Signaling Class 2 Transformer is a 120V AC input, 14V AC output transformer with a maximum load of 40VA. It has an electrostatic shield between primary and secondary windings to minimize line noise interference. This general purpose transformer steps down 120V AC to 14V AC for powering low voltage devices. It has a steel enclosure with knockouts and is UL listed. The transformer measures 5 x 4 x 3 inches and weighs 4 pounds.