Learn More
Grainger Cls 2 Trnsfrmr 5VA Enclosed Foot
Supplier:  Grainger 14W012
Encompass_Preferred
The Edwards Signaling Class 2 Transformer is a 5 VA power supply that converts 120 VAC to 24 VAC. It has a plastic enclosure and is UL listed for fire protective signaling systems. The transformer has a resettable thermal protector, terminal screws for easy wiring, and mounting tabs for installation. This transformer can provide power for up to 4 notification appliances in a notification circuit. With a compact design, it's well suited for applications where space is limited. The transformer measures just 2-3/4" x 2-1/4" x 2-3/4" (H x W x D).
Catalog No. 50-274-9724