Learn More
Cayman Chemical CLS-MPEG 2000 100mg
SDP
Catalog No. 502949890
Encompass_Preferred
A PEGylated form of cholesterol; has been used in the synthesis of liposomes for targeted drug delivery
Catalog No. 50-294-9890 Supplier Cayman Chemical Supplier No. 34647100 MG
A PEGylated form of cholesterol; has been used in the synthesis of liposomes for targeted drug delivery