Learn More
Cayman Chemical CLS-MPEG 2000 5mg
SDP
Catalog No. 502950261
Encompass_Preferred
A PEGylated form of cholesterol; has been used in the synthesis of liposomes for targeted drug delivery
Catalog No. 50-295-0261 Supplier Cayman Chemical Supplier No. 346475 MG
A PEGylated form of cholesterol; has been used in the synthesis of liposomes for targeted drug delivery