Learn More
Chemglass Life Sciences Luer Cap, Female, Non-Vented, Without Stem
SDP
Catalog No. NC2159577
Encompass
Thepolycarbonate non-vented female luer thread cover with no stem is ideal to be used as a protective cover for the Swabable Valve (CLS-1390-21V).Cap has a standard luer thread per ISO 594.For sterile applications it can be autoclaved.
This item is not returnable. View return policy
Catalog No. NC2159577 Supplier Chemglass Life Sciences Supplier No. CLS139021C
Thepolycarbonate non-vented female luer thread cover with no stem is ideal to be used as a protective cover for the Swabable Valve (CLS-1390-21V).Cap has a standard luer thread per ISO 594.For sterile applications it can be autoclaved.