Learn More
Cole-Parmer Thermo Scientific B7800-12 Vial Kit, Clear Glass, Polypropylene Cap with PTFE Liner, 12 mL; 200/Pk
Catalog No. NC2081093 Shop All Cole-Parmer Products
Encompass_Preferred
Screw-thread sample vials are manufactured from either clear 33 expansion borosilicate clear glass (Type 1, Class A) or 51A amber glass. Vials maintain PH level and eliminate ion leaching. Choose from a polypropylene cap with a PTFE-lined foam backing or a polypropylene cap with a PTFE-lined silicone septa. Vials, caps, and septa are shrink-wrapped separately.
Catalog No. NC2081093 Supplier Cole-Parmer Supplier No. 0892224
Screw-thread sample vials are manufactured from either clear 33 expansion borosilicate clear glass (Type 1, Class A) or 51A amber glass. Vials maintain PH level and eliminate ion leaching. Choose from a polypropylene cap with a PTFE-lined foam backing or a polypropylene cap with a PTFE-lined silicone septa. Vials, caps, and septa are shrink-wrapped separately.