Learn More
Lifeline Cell Technology CLSTN ALCBLU CART LIFELINE KT
SDP
Catalog No. 50311913
Encompass_Preferred
Lifeline's Alcian Blue Staining Kit contains fixative solution, sucrose stabilizer solution, Alcian Blue stain solution and acetic acid wash solution for staining sulfated proteoglycans deposited by chondrocytes.
Catalog No. 50-311-913 Supplier Lifeline Cell Technology Supplier No. LL0051
Lifeline's Alcian Blue Staining Kit contains fixative solution, sucrose stabilizer solution, Alcian Blue stain solution and acetic acid wash solution for staining sulfated proteoglycans deposited by chondrocytes.