Learn More
Lifeline Cell Technology CLSTN OILRO ADIP LIFELINE KT
SDP
Catalog No. 50311914
Encompass_Preferred
Lifeline's Oil Red O Staining Kit contains fixative solution, dehydration solution, Oil Red O stain solution and stain differential solution for staining lipid vesicles in adipocytes.
Catalog No. 50-311-914 Supplier Lifeline Cell Technology Supplier No. LL0052
Lifeline's Oil Red O Staining Kit contains fixative solution, dehydration solution, Oil Red O stain solution and stain differential solution for staining lipid vesicles in adipocytes.