Learn More
Aviva Systems Biology Corp CLSTN1 Antibody - N-terminal region (ARP44580_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP44580_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against CLSTN1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human CLSTN1. Alternate names: CSTN1, FLJ32258, KIAA0911, PIK3CD, XB31alpha, alcalpha1, alcalpha2, CDHR12, ALC-ALPHAApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-53987